0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMN CHIRALPAK IA-10 150 x 2.1 mm 10 micron SFC 1 * 1 items
Catalog # DAIC80G94
Supplier:
CHIRAL TECHNOLOGIES
Expand image
COLUMN CHIRALPAK IA-10 150 x 2.1 mm 10 micron SFC 1 * 1 items
Catalog # DAIC80G94
Supplier:
CHIRAL TECHNOLOGIES