0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMN CHIRALPAK IE 300 x 4.6 mm 20 micron 1 * 1 items
Catalog # DAIC85226
Supplier:
CHIRAL TECHNOLOGIES
Expand image
COLUMN CHIRALPAK IE 300 x 4.6 mm 20 micron 1 * 1 items
Catalog # DAIC85226
Supplier:
CHIRAL TECHNOLOGIES