Skip to Footer
Avancerad
Avancerad
SDS
Certifikat
Tillverkardata
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMN CHIRALPAK IK 50 x 4.6 mm 5 micron 1 * 1 items
Catalog # DAIC91322
Supplier:
CHIRAL TECHNOLOGIES
Expand image
COLUMN CHIRALPAK IK 50 x 4.6 mm 5 micron 1 * 1 items
Catalog # DAIC91322
Supplier:
CHIRAL TECHNOLOGIES