Order Entry
Sweden
ContactUsLinkComponent
PE-Labeled Human GUCY2C, His Tag (GUC-HP2E3) is produced via site-specific conjugation of PE to Human EGFRvIII (24-430), His Tag un der optimal conditions with a proprietary technology.PE-Labeled Human GUCY2C, His Tag (GUC-HP2E3) is expressed from hu 1 * 200 Tests
Catalog # ACRBGUC-HP2E3-200
Leverantör:  ACRO Biosystems
PE-Labeled Human GUCY2C, His Tag (GUC-HP2E3) is produced via site-specific conjugation of PE to Human EGFRvIII (24-430), His Tag un der optimal conditions with a proprietary technology.PE-Labeled Human GUCY2C, His Tag (GUC-HP2E3) is expressed from hu 1 * 200 Tests
Catalog # ACRBGUC-HP2E3-200
Leverantör:  ACRO Biosystems
Frequently Bought Together