Order Entry
Sweden
ContactUsLinkComponent
FITC-Labeled Human NCAM-1, Fc Tag (NC1-HF256) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2). 1 * 25 µG
Catalog # ACRBNC1-HF256-25UG
Supplier:  ACRO Biosystems
FITC-Labeled Human NCAM-1, Fc Tag (NC1-HF256) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2). 1 * 25 µG
Catalog # ACRBNC1-HF256-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together