Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SUN BIOSCIENCE 96 hydrogel arrays - Ø 600 - imaging bottom 1 * 1 ST
Catalog # MSPPGRI3D96IBIS966
Supplier:
MARKET SOURCE PART PROCESS
Expand image
SUN BIOSCIENCE 96 hydrogel arrays - Ø 600 - imaging bottom 1 * 1 ST
Catalog # MSPPGRI3D96IBIS966
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Mitomycin C
Supplier: Thermo Fisher Scientific
Sontara EC White, 305x380mm multipurpose nonwoven wipe: Strong, durable, absorbent, low-linting, 1 roll x 400 wipes. 1 * 1 Roll
Supplier: SONTARA
pH electrodes, InLab® ISM
Supplier: Mettler - Toledo