0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Compact Pt1000 temperature probe with silicone handle null
Catalog # GRSG609549
Supplier:
Greisinger
Expand image
Compact Pt1000 temperature probe with silicone handle null
Catalog # GRSG609549
Supplier:
Greisinger