Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Fusidate sodium

Bottle, 10 g
Catalog # APOSBIF6401-10G
Irritant
Supplier:   Apollo Scientific
Fusidate sodium
Catalog # APOSBIF6401-10G
Supplier:  Apollo Scientific
CAS Number:  751-94-0
Fusidate sodium

Bottle, 10 g

CAS Number:  751-94-0
Price
5 975,00 kr
Log in for availability
1 * 10 g
Product Details & Documents

Suppresses nitric oxide lysis of pancreatic islet cells.

Synonyms:  Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate
  • CAS Number:751-94-0
  • Formula:C₃₁H₄₇NaO₆
  • MDL Number:MFCD09054714
  • Molecular Weight:538.701 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.