Order Entry
Sweden
Orders LinkContactUsLinkComponent
Fusidate sodium

Fusidate sodium

Bottle, 5 g
Catalog # APOSBIF6401-5G
Irritant
Supplier:   Apollo Scientific
Fusidate sodium
Catalog # APOSBIF6401-5G
Supplier:  Apollo Scientific
CAS Number:  751-94-0
Fusidate sodium

Bottle, 5 g

CAS Number:  751-94-0
Price
3 750,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Suppresses nitric oxide lysis of pancreatic islet cells.

Synonyms:  Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate
  • CAS Number:751-94-0
  • Formula:C₃₁H₄₇NaO₆
  • MDL Number:MFCD09054714
  • Molecular Weight:538.701 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications