Fusidate sodium
Bottle, 1 g
Catalog # APOSBIF6401-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Suppresses nitric oxide lysis of pancreatic islet cells.
Synonyms: Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate
- CAS Number:751-94-0
- Formula:C₃₁H₄₇NaO₆
- MDL Number:MFCD09054714
- Molecular Weight:538.701 g/mol
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



