Order Entry
Sweden
ContactUsLinkComponent
FIAS Sampling Probe Assembly, 0.6mm i.d. 3.070,00 3.070,00 Includes corrosion-resistant sampling probe and screw fitting, B300-1769, and FIAS capillary tubing assembly, B300-0158. The probe is suitable for inorganic acids and most organic solvents ex 1 * 1 items
Catalog # PERKB3001771
Supplier:  PerkinElmer
FIAS Sampling Probe Assembly, 0.6mm i.d. 3.070,00 3.070,00 Includes corrosion-resistant sampling probe and screw fitting, B300-1769, and FIAS capillary tubing assembly, B300-0158. The probe is suitable for inorganic acids and most organic solvents ex 1 * 1 items
Catalog # PERKB3001771
Supplier:  PerkinElmer
Frequently Bought Together