Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Rubella virus E1 protein Fc Chimera 1 * 100 µG
Catalog # ABCAAB256429-100
Supplier:
Abcam
Expand image
Recombinant Rubella virus E1 protein Fc Chimera 1 * 100 µG
Catalog # ABCAAB256429-100
Supplier:
Abcam