0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Recombinant human MAPK6ERK3 protein 1 * 10 µG
ABCAAB217835-10
:
Abcam
Recombinant human MAPK6ERK3 protein 1 * 10 µG
ABCAAB217835-10
:
Abcam