Actarit 98%
Bottle, 5 g
Catalog # APOSOR911806-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid
- CAS Number:18699-02-0
- Formula:C₁₀H₁₁NO₃
- MDL Number:MFCD00196165
- Molecular Weight:193.20 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
- Cat. No.:APOSOR911806-5G



