Actarit 98%
Bottle, 100 g
Catalog # APOSOR911806-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid
- CAS Number:18699-02-0
- Formula:C₁₀H₁₁NO₃
- MDL Number:MFCD00196165
- Molecular Weight:193.20 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



