Order Entry
Sweden
ContactUsLinkComponent
The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike S1 protein to streptavidin conjugated magnetic beads, whi ch can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. null
Catalog # ACRBMBS-K001-10MG
Supplier:  ACRO Biosystems
The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike S1 protein to streptavidin conjugated magnetic beads, whi ch can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. null
Catalog # ACRBMBS-K001-10MG
Supplier:  ACRO Biosystems
Frequently Bought Together