Order Entry
Sweden
ContactUsLinkComponent
The biotinylated CD3E CD3D protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can captur e the anti-CD3E CD3D antibody from various assay systems. The beads are in uniform size, narrow size distribution wit 1 * 2 mg
Catalog # ACRBMBS-K003-2MG
Supplier:  ACRO Biosystems
The biotinylated CD3E CD3D protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can captur e the anti-CD3E CD3D antibody from various assay systems. The beads are in uniform size, narrow size distribution wit 1 * 2 mg
Catalog # ACRBMBS-K003-2MG
Supplier:  ACRO Biosystems
Frequently Bought Together