The biotinylated CD3E CD3D protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can captur e the anti-CD3E CD3D antibody from various assay systems. The beads are in uniform size, narrow size distribution wit 1 * 10 mg
Catalog # ACRBMBS-K003-10MG
Leverantör: ACRO Biosystems
Frequently Bought Together