The biotinylated HER2 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-HER2 antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 2 mg
Catalog # ACRBMBS-K006-2MG
Supplier: ACRO Biosystems
Frequently Bought Together