Order Entry
Sweden
ContactUsLinkComponent
The biotinylated CD38 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-CD38 antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 10 mg
  ACRBMBC-K010-10MG
The biotinylated CD38 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-CD38 antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 10 mg
  ACRBMBC-K010-10MG
Frequently Bought Together