Order Entry
Sweden
ContactUsLinkComponent
SARS-CoV-2 S protein RBD, Mouse IgG2a Fc TagMALS verified SPD-C5259 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accession QHD43416.1. 1 * 100 µG
Catalog # ACRBSPD-C5259100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD, Mouse IgG2a Fc TagMALS verified SPD-C5259 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accession QHD43416.1. 1 * 100 µG
Catalog # ACRBSPD-C5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together