Order Entry
Sweden
ContactUsLinkComponent
The streptavidin magnetic beads are uniform size, narrow size distribution with large surface area and unique surface coating, it is easy to capture the biotinylated proteins or other molecules, and the bounded protein have no activity lost, can hel 1 * 5 mg
Catalog # ACRBSMB-B01-5MG
Supplier:  ACRO Biosystems
The streptavidin magnetic beads are uniform size, narrow size distribution with large surface area and unique surface coating, it is easy to capture the biotinylated proteins or other molecules, and the bounded protein have no activity lost, can hel 1 * 5 mg
Catalog # ACRBSMB-B01-5MG
Supplier:  ACRO Biosystems
Frequently Bought Together