Order Entry
Sweden
ContactUsLinkComponent
Biotinylated Human PD-1, Fc,Avitag,His Tag PD1-H82F4 is expressed from human 293 cells HEK293. It contains AA Leu 25 - Gln 167 Acces sion Q15116-1. 1 * 200 µG
Catalog # ACRBPD1-H82F4200UG
Supplier:  ACRO Biosystems
Biotinylated Human PD-1, Fc,Avitag,His Tag PD1-H82F4 is expressed from human 293 cells HEK293. It contains AA Leu 25 - Gln 167 Acces sion Q15116-1. 1 * 200 µG
Catalog # ACRBPD1-H82F4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together