Order Entry
Sweden
ContactUsLinkComponent
Biotinylated SARS-CoV-2 S protein RBD, Mouse IgG1 Fc,Avitag™MALS verified SPD-C82Aa is expressed from human 293 cells HEK293. It con tains AA Arg 319 - Lys 537 Accession QHD43416.1. 1 * 25 µG
Catalog # ACRBSPD-C82AA-25UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 S protein RBD, Mouse IgG1 Fc,Avitag™MALS verified SPD-C82Aa is expressed from human 293 cells HEK293. It con tains AA Arg 319 - Lys 537 Accession QHD43416.1. 1 * 25 µG
Catalog # ACRBSPD-C82AA-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together