Order Entry
Sweden
ContactUsLinkComponent
The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike trimer protein to streptavidin conjugated magnetic beads, which can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. The SARS-CoV-2 Spike trim 1 * 2 mg
Catalog # ACRBMBS-K015-2MG
Supplier:  ACRO Biosystems
The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike trimer protein to streptavidin conjugated magnetic beads, which can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. The SARS-CoV-2 Spike trim 1 * 2 mg
Catalog # ACRBMBS-K015-2MG
Supplier:  ACRO Biosystems
Frequently Bought Together