Order Entry
Sweden
ContactUsLinkComponent
Cynomolgus CD3ECD3D Heterodimer Protein, Fc,His TagFc,Flag Tag CDD-C52W9 is expressed from human 293 cells HEK293. It contains AA Gl n 22 - Asp 117 CD3E Phe 22 - Ala 105 CD3D Accession Q95LI5-1 CD3E Q95LI8-1 CD3D. 1 * 100 µG
Catalog # ACRBCDD-C52W9100UG
Supplier:  ACRO Biosystems
Cynomolgus CD3ECD3D Heterodimer Protein, Fc,His TagFc,Flag Tag CDD-C52W9 is expressed from human 293 cells HEK293. It contains AA Gl n 22 - Asp 117 CD3E Phe 22 - Ala 105 CD3D Accession Q95LI5-1 CD3E Q95LI8-1 CD3D. 1 * 100 µG
Catalog # ACRBCDD-C52W9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together