Order Entry
Sweden
ContactUsLinkComponent
Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Human IgG1 AM128 is isolated from a SARS-CoV-2 infected patient and is recombinant ly produced from human 293 cells HEK293. This antibody recognizes the SARS-CoV-2 Spike Protein RBD domain and inhibit 1 * 1 mg
Catalog # ACRBSPD-M180-1MG
Supplier:  ACROBIOSYSTEMS
Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Human IgG1 AM128 is isolated from a SARS-CoV-2 infected patient and is recombinant ly produced from human 293 cells HEK293. This antibody recognizes the SARS-CoV-2 Spike Protein RBD domain and inhibit 1 * 1 mg
Catalog # ACRBSPD-M180-1MG
Supplier:  ACROBIOSYSTEMS