Order Entry
Sweden
ContactUsLinkComponent
Glove R068 R-Flex seamless HPPE knit featuring Nitrile Full Dip with cut protection, high dexterity and liquid resistance. Touch Screen compatible on first two fingers and thumb. Size 7 1 * 12 PAIR
Catalog # ANSER068070
Supplier:  Ansell
Glove R068 R-Flex seamless HPPE knit featuring Nitrile Full Dip with cut protection, high dexterity and liquid resistance. Touch Screen compatible on first two fingers and thumb. Size 7 1 * 12 PAIR
Catalog # ANSER068070
Supplier:  Ansell