Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Solubility inserts ADPI, 8 sleeves made of stainless steel and 8 reduction rings 1 * 8 Items
Catalog # GEAG03.4102
Supplier:
Gerber Instruments AG
Expand image
Solubility inserts ADPI, 8 sleeves made of stainless steel and 8 reduction rings 1 * 8 Items
Catalog # GEAG03.4102
Supplier:
Gerber Instruments AG