Order Entry
Sweden
ContactUsLinkComponent
Purified recombinant protein of Human cytochrome P450, family 19, subfamily A, polypeptide 1 (CYP19A1), transcript variant 1, full length, with C-terminal MYC/DDK tag, expressed in HEK293 cells 1 * 20 µG
Catalog # ORIGTP305890
Supplier:  OriGene
Purified recombinant protein of Human cytochrome P450, family 19, subfamily A, polypeptide 1 (CYP19A1), transcript variant 1, full length, with C-terminal MYC/DDK tag, expressed in HEK293 cells 1 * 20 µG
Catalog # ORIGTP305890
Supplier:  OriGene
Frequently Bought Together