0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COSMOSIL(R) Cholester Packed Column 20mmI.D.x250mm 1 * 1 items
Catalog # HISM05982-71
Supplier:
HICHROM
Expand image
COSMOSIL(R) Cholester Packed Column 20mmI.D.x250mm 1 * 1 items
Catalog # HISM05982-71
Supplier:
HICHROM