Order Entry
Sweden
ContactUsLinkComponent
Recirculating Cooler Unichiller 075 with Pilot ONE -20...40 °C 1 * 1 items
  HUBE3040.0031.01
 :  HUBER
Recirculating Cooler Unichiller 075 with Pilot ONE -20...40 °C 1 * 1 items
  HUBE3040.0031.01
 :  HUBER