High performance thermometer (type K thermocouple), IP65 rated, with BioCote® antimicrobial protection, selectable C/F scales with d efault to °F, MAX/MIN, Audible and Visial Alarms, clock, 1 * 1 Items
Catalog # MSPP5070163
Supplier: MARKET SOURCE PART PROCESS