Order Entry
Sweden
ContactUsLinkComponent
Human IL-13 R alpha 1, His Tag (SPR verified) (IL1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 34 3 (Accession NP001551.1). 1 * 1 mg
Catalog # ACRBIL1-H5224-1MG
Supplier:  ACROBIOSYSTEMS
Human IL-13 R alpha 1, His Tag (SPR verified) (IL1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 34 3 (Accession NP001551.1). 1 * 1 mg
Catalog # ACRBIL1-H5224-1MG
Supplier:  ACROBIOSYSTEMS