Order Entry
Sweden
ContactUsLinkComponent
Human TSLP (R127A, R130A), Mouse IgG2a Fc Tag (TSP-H5255) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 15 9 (Accession Q969D9-1 (R127A, R130A)). 1 * 100 µG
Catalog # ACRBTSP-H5255100UG
Supplier:  ACRO Biosystems
Human TSLP (R127A, R130A), Mouse IgG2a Fc Tag (TSP-H5255) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 15 9 (Accession Q969D9-1 (R127A, R130A)). 1 * 100 µG
Catalog # ACRBTSP-H5255100UG
Supplier:  ACRO Biosystems
Frequently Bought Together