Order Entry
Sweden
ContactUsLinkComponent
Human CD3 epsilon, Fc,His Tag (CDE-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession NP0 00724.1). 1 * 100 µG
Catalog # ACRBCDE-H5256100UG
Supplier:  ACROBIOSYSTEMS
Human CD3 epsilon, Fc,His Tag (CDE-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession NP0 00724.1). 1 * 100 µG
Catalog # ACRBCDE-H5256100UG
Supplier:  ACROBIOSYSTEMS