Order Entry
Sweden
ContactUsLinkComponent
ActiveMax® Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession NP000749.2). 1 * 50 µG
  ACRBGMF-H4214-50UG
ActiveMax® Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession NP000749.2). 1 * 50 µG
  ACRBGMF-H4214-50UG
Frequently Bought Together