Order Entry
Sweden
ContactUsLinkComponent
ActiveMax® Human TGFB1, Tag Free (TG1-H4212) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession P01137-1). 1 * 50 µG
  ACRBTG1-H4212-50UG
ActiveMax® Human TGFB1, Tag Free (TG1-H4212) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession P01137-1). 1 * 50 µG
  ACRBTG1-H4212-50UG
Frequently Bought Together