Order Entry
Sweden
ContactUsLinkComponent
ActiveMax® Human VEGF110, Tag Free (HPLC-verified) (VE0-H5212) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 136 (Accession NP001165097). 1 * 500 µG
Catalog # ACRBVE0-H5212500UG
Supplier:  ACROBIOSYSTEMS
ActiveMax® Human VEGF110, Tag Free (HPLC-verified) (VE0-H5212) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 136 (Accession NP001165097). 1 * 500 µG
Catalog # ACRBVE0-H5212500UG
Supplier:  ACROBIOSYSTEMS