Order Entry
Sweden
ContactUsLinkComponent
Human ErbB3, Strep Tag (ER3-H5288) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession NP001973.2 ). 1 * 250 µG
Catalog # ACRBER3-H5288250UG
Supplier:  ACROBIOSYSTEMS
Human ErbB3, Strep Tag (ER3-H5288) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession NP001973.2 ). 1 * 250 µG
Catalog # ACRBER3-H5288250UG
Supplier:  ACROBIOSYSTEMS