Order Entry
Sweden
ContactUsLinkComponent
Recombinat Protein,Human FABP3 H-FABP Protein, His Tag,Human FABP3, His Tag (FA3-H5128) is expressed from E.coli cells. It contains AA Met 1 - Ala 133 (Accession NP004093).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBFA3-H5128-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human FABP3 H-FABP Protein, His Tag,Human FABP3, His Tag (FA3-H5128) is expressed from E.coli cells. It contains AA Met 1 - Ala 133 (Accession NP004093).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBFA3-H5128-1MG
Supplier:  ACROBIOSYSTEMS