Order Entry
Sweden
ContactUsLinkComponent
MABSol® Biotinylated Human CD47, Fc,Avitag (CD7-H82F6) is expressed from human HEK293 cells. It contains AA Gln 19 - Pro 139 (Access ion NP942088). 1 * 200 µG
Catalog # ACRBCD7-H82F6200UG
Supplier:  ACROBIOSYSTEMS
MABSol® Biotinylated Human CD47, Fc,Avitag (CD7-H82F6) is expressed from human HEK293 cells. It contains AA Gln 19 - Pro 139 (Access ion NP942088). 1 * 200 µG
Catalog # ACRBCD7-H82F6200UG
Supplier:  ACROBIOSYSTEMS