Order Entry
Sweden
ContactUsLinkComponent
Biotinylated Human 2B4, Avitag,His Tag (recommended for biopanning) (2B4-H82E9) is expressed from human 293 cells (HEK293). It conta ins AA Cys 22 - Arg 221 (Accession Q9BZW8-2). 1 * 25 µG
Catalog # ACRB2B4-H82E9-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human 2B4, Avitag,His Tag (recommended for biopanning) (2B4-H82E9) is expressed from human 293 cells (HEK293). It conta ins AA Cys 22 - Arg 221 (Accession Q9BZW8-2). 1 * 25 µG
Catalog # ACRB2B4-H82E9-25UG
Supplier:  ACROBIOSYSTEMS