Order Entry
Sweden
ContactUsLinkComponent
Human CD19 (20-291), His Tag (CD9-H52H2) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession P15 391-1). 1 * 50 µG
Catalog # ACRBCD9-H52H2-50UG
Supplier:  ACROBIOSYSTEMS
Human CD19 (20-291), His Tag (CD9-H52H2) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession P15 391-1). 1 * 50 µG
Catalog # ACRBCD9-H52H2-50UG
Supplier:  ACROBIOSYSTEMS