Order Entry
Sweden
ContactUsLinkComponent
Human ITGAEITGB7 Heterodimer Protein, His TagTag Free (IT7-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Ser 1124 (ITGAE) Glu 20 - His 723 (ITGB7) (Accession P38570-1 (ITGAE) P26010-1 (ITGB7)). 1 * 1 mg
Catalog # ACRBIT7-H52W7-1MG
Supplier:  ACRO Biosystems
Human ITGAEITGB7 Heterodimer Protein, His TagTag Free (IT7-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Ser 1124 (ITGAE) Glu 20 - His 723 (ITGB7) (Accession P38570-1 (ITGAE) P26010-1 (ITGB7)). 1 * 1 mg
Catalog # ACRBIT7-H52W7-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together