Order Entry
Sweden
ContactUsLinkComponent
Biotinylated Human CD23, His,Avitag (CD3-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accessi on P06734-1). 1 * 200 µG
  ACRBCD3-H82Q5200UG
Biotinylated Human CD23, His,Avitag (CD3-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accessi on P06734-1). 1 * 200 µG
  ACRBCD3-H82Q5200UG