Order Entry
Sweden
ContactUsLinkComponent
Human B7-2, Fc Tag (CD6-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession AAH40261). 1 * 200 µG
Catalog # ACRBCD6-H5257200UG
Supplier:  ACROBIOSYSTEMS
Human B7-2, Fc Tag (CD6-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession AAH40261). 1 * 200 µG
Catalog # ACRBCD6-H5257200UG
Supplier:  ACROBIOSYSTEMS