Order Entry
Sweden
ContactUsLinkComponent
Recombinat Protein,Human Galectin-3 Protein, His Tag,Human Galectin-3, His Tag (GA3-H5247) is expressed from human 293 cells (HEK293 ). It contains AA Met 1 - Ile 250 (Accession P17931-1).This protein carries a polyhistidine tag at the N-terminus. Th 1 * 100 µG
Catalog # ACRBGA3-H5247100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Galectin-3 Protein, His Tag,Human Galectin-3, His Tag (GA3-H5247) is expressed from human 293 cells (HEK293 ). It contains AA Met 1 - Ile 250 (Accession P17931-1).This protein carries a polyhistidine tag at the N-terminus. Th 1 * 100 µG
Catalog # ACRBGA3-H5247100UG
Supplier:  ACROBIOSYSTEMS