Order Entry
Sweden
ContactUsLinkComponent
Biotinylated Cynomolgus Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are 1 * 25 µG
Catalog # ACRBBCA-C82F4-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Cynomolgus Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are 1 * 25 µG
Catalog # ACRBBCA-C82F4-25UG
Supplier:  ACROBIOSYSTEMS