Order Entry
Sweden
ContactUsLinkComponent
Human CD38, mouse IgG2a Fc Tag, low endotoxin (CD8-H5253) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 30 0 (Accession P28907-1). 1 * 50 µG
Catalog # ACRBCD8-H5253-50UG
Supplier:  ACRO Biosystems
Human CD38, mouse IgG2a Fc Tag, low endotoxin (CD8-H5253) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 30 0 (Accession P28907-1). 1 * 50 µG
Catalog # ACRBCD8-H5253-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together